Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005911.5 | NP_005902.1 | 8 - 9 | 1394 | 110 |
GenBank mRNA | AK291126.1 | - | 8 - 9 | 1202 | 110 |
AK303877.1 | - | 3 - 4 | 364 | 110 | |
BC001686.2 | - | 8 - 9 | 1191 | 110 | |
BC001854.2 | - | 8 - 9 | 1191 | 110 | |
DQ083239.1 | - | 8 - 9 | 1130 | 110 | |
X68836.1 | - | 8 - 9 | 1149 | 110 |