Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005768.5 | NP_005759.4 | 7 - 8 | 905 | 121 |
GenBank mRNA | AK096775.1 | - | 7 - 8 | 1094 | 121 |
AK296145.1 | - | 6 - 7 | 615 | 121 | |
AK296164.1 | - | 6 - 7 | 662 | 121 | |
AK296171.1 | - | 6 - 7 | 803 | 121 | |
AK311727.1 | - | 5 - 6 | 627 | 121 | |
AK315538.1 | - | 5 - 6 | 477 | 121 | |
BC000664.2 | - | 6 - 7 | 640 | 121 | |
BC065194.1 | - | 7 - 8 | 836 | 121 | |
BT007000.1 | - | 5 - 6 | 473 | 121 | |
BX647983.1 | - | 7 - 8 | 845 | 121 | |
BX648009.1 | - | 7 - 8 | 876 | 121 | |
U72515.1 | - | 6 - 7 | 590 | 121 |