Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005768.5 | NP_005759.4 | 8 - 9 | 981 | 101 |
GenBank mRNA | AK058063.1 | - | 2 - 3 | 263 | 101 |
AK096775.1 | - | 8 - 9 | 1170 | 101 | |
AK296145.1 | - | 7 - 8 | 691 | 101 | |
AK296164.1 | - | 7 - 8 | 738 | 101 | |
AK296171.1 | - | 7 - 8 | 879 | 101 | |
AK311727.1 | - | 6 - 7 | 703 | 101 | |
AK315538.1 | - | 6 - 7 | 553 | 101 | |
BC000664.2 | - | 7 - 8 | 716 | 101 | |
BC065194.1 | - | 8 - 9 | 912 | 101 | |
BT007000.1 | - | 6 - 7 | 549 | 101 | |
BX647983.1 | - | 8 - 9 | 921 | 101 | |
BX648009.1 | - | 8 - 9 | 952 | 101 | |
U72515.1 | - | 7 - 8 | 666 | 101 | |
U72517.1 | - | 1 - 2 | 662 | 101 |