Assay Design



Gene Transcripts

Gene Symbol
SSR2
Entrez Gene ID
6746
Gene Name
signal sequence receptor subunit 2
Gene Aliases
HSD25, TLAP, TRAP-BETA, TRAPB
Chromosome Location
Chr.1: 156009048 - 156020992 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003145.3 NP_003136.1 4 - 5 445 96
XM_011509923.2 XP_011508225.1 4 - 5 470 96
GenBank mRNA AK222600.1 - 4 - 5 388 96
AK300678.1 - 4 - 5 445 96
AK313821.1 - 4 - 5 445 96
AY251536.1 - 4 - 5 405 96
BC000341.2 - 4 - 5 391 96
CR456973.1 - 3 - 4 365 96
D37991.1 - 4 - 5 389 96
X74104.1 - 4 - 5 415 96

View Full Product Details