Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004128.2 | NP_004119.1 | 5 - 6 | 551 | 86 |
XM_011535052.2 | XP_011533354.1 | 6 - 7 | 661 | 86 | |
XM_017020551.1 | XP_016876040.1 | 7 - 8 | 795 | 86 | |
GenBank mRNA | AK291545.1 | - | 5 - 6 | 544 | 86 |
BC001771.1 | - | 5 - 6 | 512 | 86 | |
BT019525.1 | - | 5 - 6 | 391 | 86 | |
X16901.1 | - | 5 - 6 | 510 | 86 | |
X59745.1 | - | 5 - 6 | 391 | 86 |