Assay Design



Gene Transcripts

Gene Symbol
ERC1
Entrez Gene ID
23085
Gene Name
ELKS/RAB6-interacting/CAST family member 1
Gene Aliases
Cast2, ELKS, ERC-1, RAB6IP2
Chromosome Location
Chr.12: 990010 - 1495933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301248.1 NP_001288177.1 15 - 16 3080 66
NM_178039.3 NP_829883.1 15 - 16 3111 66
NM_178040.3 NP_829884.1 16 - 17 3195 66
NR_027946.2 15 - 16 3156 66
NR_027948.2 16 - 17 3288 66
NR_027949.2 12 - 13 2307 66
XM_011520941.2 XP_011519243.1 16 - 17 3239 66
XM_017019052.1 XP_016874541.1 18 - 19 3380 66
XM_017019053.1 XP_016874542.1 18 - 19 3314 66
XM_017019054.1 XP_016874543.1 18 - 19 3240 66
XM_017019055.1 XP_016874544.1 18 - 19 3236 66
XM_017019056.1 XP_016874545.1 18 - 19 3177 66
XM_017019057.1 XP_016874546.1 18 - 19 3371 66
XM_017019058.1 XP_016874547.1 18 - 19 3305 66
XM_017019059.1 XP_016874548.1 17 - 18 3368 66
XM_017019060.1 XP_016874549.1 17 - 18 3359 66
XM_017019061.1 XP_016874550.1 17 - 18 3326 66
XM_017019062.1 XP_016874551.1 17 - 18 3317 66
XM_017019063.1 XP_016874552.1 16 - 17 3314 66
XM_017019064.1 XP_016874553.1 16 - 17 3305 66
XM_017019065.1 XP_016874554.1 17 - 18 3287 66
XM_017019066.1 XP_016874555.1 16 - 17 3233 66
XM_017019067.1 XP_016874556.1 15 - 16 3221 66
XM_017019068.1 XP_016874557.1 16 - 17 3305 66
XM_017019069.1 XP_016874558.1 16 - 17 3194 66
XM_017019070.1 XP_016874559.1 15 - 16 3173 66
XM_017019071.1 XP_016874560.1 15 - 16 3101 66
XM_017019072.1 XP_016874561.1 18 - 19 3380 66
XM_017019074.1 XP_016874563.1 11 - 12 1360 66
XM_017019075.1 XP_016874564.1 10 - 11 1304 66
GenBank mRNA AB015617.1 - 15 - 16 3114 66
AB029004.2 - 15 - 16 2975 66
AB053468.1 - 16 - 17 3229 66
AB053469.1 - 12 - 13 2242 66
AB053470.1 - 15 - 16 3053 66
AB053471.1 - 16 - 17 3105 66
AK000148.1 - 3 - 4 445 66
AK092201.1 - 5 - 6 730 66
AK294351.1 - 13 - 14 2075 66
BC037377.1 - 5 - 6 718 66
BC132782.1 - 15 - 16 3056 66
BC132784.1 - 15 - 16 3056 66
BC144287.1 - 15 - 16 3056 66
BC150248.1 - 15 - 16 2996 66

View Full Product Details