Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032656.3 | NP_116045.2 | 23 - 24 | 3220 | 90 |
XM_005253590.3 | XP_005253647.1 | 23 - 24 | 3224 | 90 | |
GenBank mRNA | AB040950.1 | - | 20 - 21 | 2592 | 90 |
AK123968.1 | - | 2 - 3 | 1429 | 90 | |
BC002575.2 | - | 20 - 21 | 2603 | 90 | |
BC037964.1 | - | 23 - 24 | 3202 | 90 |