Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032656.3 | NP_116045.2 | 5 - 6 | 992 | 107 |
XM_005253590.3 | XP_005253647.1 | 5 - 6 | 996 | 107 | |
XM_011538598.2 | XP_011536900.1 | 5 - 6 | 996 | 107 | |
XM_011538600.2 | XP_011536902.1 | 5 - 6 | 996 | 107 | |
GenBank mRNA | AB040950.1 | - | 2 - 3 | 364 | 107 |
BC002575.2 | - | 2 - 3 | 375 | 107 | |
BC037964.1 | - | 5 - 6 | 974 | 107 |