Assay Design



Gene Transcripts

Gene Symbol
IFT81
Entrez Gene ID
28981
Gene Name
intraflagellar transport 81
Gene Aliases
CDV-1, CDV-1R, CDV1, CDV1R, DV1
Chromosome Location
Chr.12: 110124335 - 110218795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143779.1 NP_001137251.1 6 - 7 722 111
NM_014055.3 NP_054774.2 6 - 7 1140 111
NM_031473.3 NP_113661.2 6 - 7 1140 111
XM_011538232.2 XP_011536534.1 6 - 7 668 111
XM_017019217.1 XP_016874706.1 6 - 7 832 111
XM_017019218.1 XP_016874707.1 6 - 7 1140 111
GenBank mRNA AF139540.1 - 6 - 7 680 111
AF250326.1 - 5 - 6 633 111
AF332010.1 - 6 - 7 1098 111
AK091549.1 - 5 - 6 500 111
AK291043.1 - 6 - 7 1042 111
BC004536.2 - 6 - 7 976 111
BC005396.1 - 6 - 7 642 111
BC108257.1 - 5 - 6 605 111

View Full Product Details