Assay Design



Gene Transcripts

Gene Symbol
DGCR14
Entrez Gene ID
8220
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
DGCR13, DGS-H, DGS-I, DGSH, DGSI, ES2, Es2el
Chromosome Location
Chr.22: 19130279 - 19144726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022719.2 NP_073210.1 7 - 8 956 91
XM_005261282.4 XP_005261339.1 7 - 8 964 91
XM_006724329.3 XP_006724392.1 7 - 8 1323 91
XM_006724330.3 XP_006724393.1 7 - 8 1364 91
XM_006724331.3 XP_006724394.1 7 - 8 1361 91
XM_017028958.1 XP_016884447.1 7 - 8 1320 91
GenBank mRNA BC003015.1 - 7 - 8 918 91
BC006542.2 - 7 - 8 920 91
CR456344.1 - 7 - 8 928 91
L77566.1 - 7 - 8 929 91
L78010.1 - 7 - 8 925 91

View Full Product Details