Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001670.2 | NP_001661.1 | 2 - 3 | 256 | 68 |
XM_005261242.2 | XP_005261299.1 | 2 - 3 | 272 | 68 | |
XM_005261243.3 | XP_005261300.1 | 2 - 3 | 272 | 68 | |
XM_005261244.3 | XP_005261301.1 | 2 - 3 | 272 | 68 | |
XM_006724243.2 | XP_006724306.1 | 2 - 3 | 272 | 68 | |
XM_006724245.2 | XP_006724308.1 | 2 - 3 | 272 | 68 | |
XM_006724249.3 | XP_006724312.1 | 2 - 3 | 272 | 68 | |
XM_006724250.3 | XP_006724313.1 | 2 - 3 | 272 | 68 | |
XM_011530179.2 | XP_011528481.1 | 2 - 3 | 272 | 68 | |
XM_011530180.1 | XP_011528482.1 | 2 - 3 | 272 | 68 | |
XM_011530181.1 | XP_011528483.1 | 2 - 3 | 272 | 68 | |
GenBank mRNA | U51269.1 | - | 2 - 3 | 257 | 68 |