Assay Design



Gene Transcripts

Gene Symbol
ARVCF
Entrez Gene ID
421
Gene Name
armadillo repeat gene deleted in velocardiofacial syndrome
Gene Aliases
-
Chromosome Location
Chr.22: 19966491 - 20016802 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001670.2 NP_001661.1 4 - 5 635 95
XM_005261242.2 XP_005261299.1 4 - 5 651 95
XM_005261243.3 XP_005261300.1 4 - 5 651 95
XM_005261244.3 XP_005261301.1 4 - 5 651 95
XM_006724243.2 XP_006724306.1 4 - 5 651 95
XM_006724245.2 XP_006724308.1 4 - 5 651 95
XM_006724246.3 XP_006724309.1 2 - 3 3487 95
XM_006724247.3 XP_006724310.1 2 - 3 295 95
XM_006724248.3 XP_006724311.1 2 - 3 294 95
XM_006724249.3 XP_006724312.1 4 - 5 651 95
XM_006724250.3 XP_006724313.1 4 - 5 651 95
XM_011530179.2 XP_011528481.1 4 - 5 651 95
XM_011530180.1 XP_011528482.1 4 - 5 651 95
XM_011530181.1 XP_011528483.1 4 - 5 651 95
GenBank mRNA BC036893.1 - 3 - 4 415 95
BC137436.1 - 3 - 4 413 95
U51269.1 - 4 - 5 636 95

View Full Product Details