Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001670.2 | NP_001661.1 | 9 - 10 | 2137 | 74 |
XM_006724243.2 | XP_006724306.1 | 9 - 10 | 2153 | 74 | |
XM_006724245.2 | XP_006724308.1 | 9 - 10 | 2153 | 74 | |
XM_006724246.3 | XP_006724309.1 | 7 - 8 | 4989 | 74 | |
XM_006724247.3 | XP_006724310.1 | 7 - 8 | 1797 | 74 | |
XM_006724249.3 | XP_006724312.1 | 9 - 10 | 2153 | 74 | |
XM_011530180.1 | XP_011528482.1 | 9 - 10 | 2153 | 74 | |
XM_011530182.2 | XP_011528484.1 | 4 - 5 | 765 | 74 | |
GenBank mRNA | BC137436.1 | - | 8 - 9 | 1915 | 74 |
U51269.1 | - | 9 - 10 | 2138 | 74 |