Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099274.1 | NP_001092744.1 | 1 - 2 | 539 | 57 |
NM_012461.2 | NP_036593.2 | 1 - 2 | 539 | 57 | |
XM_011536642.2 | XP_011534944.1 | 1 - 2 | 511 | 57 | |
GenBank mRNA | AF195512.1 | - | 1 - 2 | 459 | 57 |
AK225379.1 | - | 1 - 2 | 483 | 57 | |
AK315084.1 | - | 1 - 2 | 427 | 57 | |
BC005030.1 | - | 1 - 2 | 320 | 57 | |
BC019343.1 | - | 1 - 2 | 497 | 57 | |
BM549897.1 | - | 1 - 2 | 522 | 57 | |
BX161478.1 | - | 1 - 2 | 472 | 57 | |
DA053760.1 | - | 1 - 2 | 539 | 57 |