Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001131005.2 | NP_001124477.1 | 8 - 9 | 1228 | 62 |
NM_001193347.1 | NP_001180276.1 | 9 - 10 | 1288 | 62 | |
NM_001193348.1 | NP_001180277.1 | 5 - 6 | 800 | 62 | |
NM_001308002.1 | NP_001294931.1 | 6 - 7 | 944 | 62 | |
XM_006714625.3 | XP_006714688.1 | 8 - 9 | 1346 | 62 | |
XM_011543401.1 | XP_011541703.1 | 7 - 8 | 1231 | 62 | |
XM_017009476.1 | XP_016864965.1 | 7 - 8 | 1292 | 62 | |
XM_017009477.1 | XP_016864966.1 | 7 - 8 | 1231 | 62 | |
XM_017009478.1 | XP_016864967.1 | 7 - 8 | 1286 | 62 | |
XM_017009482.1 | XP_016864971.1 | 7 - 8 | 1292 | 62 | |
XM_017009483.1 | XP_016864972.1 | 7 - 8 | 1286 | 62 | |
GenBank mRNA | AL833268.1 | - | 9 - 10 | 1236 | 62 |
AL833274.1 | - | 8 - 9 | 1177 | 62 | |
FM163484.2 | - | 5 - 6 | 661 | 62 | |
S57212.1 | - | - | 1465 | 62 |