Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130853.2 | NP_570923.2 | 6 - 7 | 913 | 70 |
NM_130854.2 | NP_570924.2 | 6 - 7 | 913 | 70 | |
XM_005259600.2 | XP_005259657.1 | 6 - 7 | 881 | 70 | |
XM_005259606.2 | XP_005259663.1 | 6 - 7 | 881 | 70 | |
XM_005259607.2 | XP_005259664.1 | 6 - 7 | 881 | 70 | |
XM_005259609.1 | XP_005259666.1 | 6 - 7 | 880 | 70 | |
XM_005259610.1 | XP_005259667.1 | 6 - 7 | 880 | 70 | |
XM_011528157.2 | XP_011526459.1 | 5 - 6 | 751 | 70 | |
GenBank mRNA | AB209333.1 | - | 6 - 7 | 913 | 70 |
BC104812.1 | - | 5 - 6 | 753 | 70 | |
BC143286.1 | - | 5 - 6 | 753 | 70 | |
BC143287.1 | - | 5 - 6 | 753 | 70 | |
U40317.1 | - | 5 - 6 | 701 | 70 | |
U41725.1 | - | 5 - 6 | 701 | 70 |