Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005455.4 | NP_005446.2 | 2 - 3 | 398 | 106 |
NM_203350.2 | NP_976225.1 | 2 - 3 | 398 | 106 | |
GenBank mRNA | AB209568.1 | - | 2 - 3 | 111 | 106 |
AF065391.1 | - | 2 - 3 | 138 | 106 | |
AF065392.1 | - | 2 - 3 | 138 | 106 | |
AL136945.1 | - | 2 - 3 | 120 | 106 | |
AU100296.1 | - | 2 - 3 | 172 | 106 | |
BC033648.1 | - | 2 - 3 | 144 | 106 | |
BC039814.1 | - | 2 - 3 | 144 | 106 |