Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000056.4 | NP_000047.1 | 1 - 2 | 279 | 87 |
NM_183050.3 | NP_898871.1 | 1 - 2 | 279 | 87 | |
NR_134945.1 | 1 - 2 | 279 | 87 | ||
XM_005248756.4 | XP_005248813.1 | 1 - 2 | 228 | 87 | |
XM_011536023.2 | XP_011534325.1 | 1 - 2 | 228 | 87 | |
XM_011536024.2 | XP_011534326.1 | 1 - 2 | 228 | 87 | |
XM_011536025.2 | XP_011534327.1 | 1 - 2 | 228 | 87 | |
GenBank mRNA | AK289977.1 | - | 1 - 2 | 208 | 87 |
AK316545.1 | - | 2 - 3 | 192 | 87 | |
BC034481.1 | - | 1 - 2 | 223 | 87 | |
BC040139.2 | - | 1 - 2 | 200 | 87 | |
BT020063.1 | - | 1 - 2 | 195 | 87 | |
DC372349.1 | - | 1 - 2 | 267 | 87 | |
DC422376.1 | - | 1 - 2 | 282 | 87 | |
M55575.1 | - | 1 - 2 | 199 | 87 | |
U50708.1 | - | 1 - 2 | 242 | 87 | |
X52446.1 | - | 1 - 2 | 138 | 87 |