Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005458.7 | NP_005449.5 | 7 - 8 | 1699 | 63 |
XM_005252316.4 | XP_005252373.1 | 5 - 6 | 732 | 63 | |
XM_017015331.1 | XP_016870820.1 | 6 - 7 | 1117 | 63 | |
XM_017015332.1 | XP_016870821.1 | 4 - 5 | 686 | 63 | |
GenBank mRNA | AF056085.1 | - | 7 - 8 | 1696 | 63 |
AF069755.1 | - | 7 - 8 | 1395 | 63 | |
AF074483.1 | - | 7 - 8 | 1239 | 63 | |
AF095784.1 | - | 7 - 8 | 1370 | 63 | |
AF099033.1 | - | 7 - 8 | 1239 | 63 | |
AJ012188.1 | - | 7 - 8 | 1239 | 63 | |
AK308139.1 | - | 6 - 7 | 872 | 63 | |
BC035071.2 | - | 8 - 9 | 1534 | 63 | |
JQ308173.1 | - | 7 - 8 | 1239 | 63 |