Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005458.7 | NP_005449.5 | 8 - 9 | 1760 | 95 |
XM_005252316.4 | XP_005252373.1 | 6 - 7 | 793 | 95 | |
XM_017015331.1 | XP_016870820.1 | 7 - 8 | 1178 | 95 | |
XM_017015332.1 | XP_016870821.1 | 5 - 6 | 747 | 95 | |
GenBank mRNA | AF056085.1 | - | 8 - 9 | 1757 | 95 |
AF069755.1 | - | 8 - 9 | 1456 | 95 | |
AF074483.1 | - | 8 - 9 | 1300 | 95 | |
AF095784.1 | - | 8 - 9 | 1431 | 95 | |
AF099033.1 | - | 8 - 9 | 1300 | 95 | |
AJ012188.1 | - | 8 - 9 | 1300 | 95 | |
AK308139.1 | - | 7 - 8 | 933 | 95 | |
BC035071.2 | - | 9 - 10 | 1595 | 95 | |
JQ308173.1 | - | 8 - 9 | 1300 | 95 |