Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006454.2 | NP_006445.1 | 4 - 5 | 342 | 73 |
XM_011513388.2 | XP_011511690.1 | 5 - 6 | 847 | 73 | |
GenBank mRNA | AF040963.1 | - | 4 - 5 | 323 | 73 |
AF082740.1 | - | 4 - 5 | 342 | 73 | |
AF187724.1 | - | 3 - 4 | 179 | 73 | |
AK024501.1 | - | 1 - 2 | 2828 | 73 | |
BC068060.1 | - | 4 - 5 | 395 | 73 | |
BT019969.1 | - | 4 - 5 | 310 | 73 | |
BT019970.1 | - | 4 - 5 | 310 | 73 |