Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019098.4 | NP_061971.3 | 8 - 9 | 1039 | 147 |
XM_011517138.2 | XP_011515440.1 | 6 - 7 | 694 | 147 | |
GenBank mRNA | AF228520.1 | - | 5 - 6 | 652 | 147 |
AF272900.1 | - | 8 - 9 | 1037 | 147 | |
BC150601.1 | - | 8 - 9 | 1017 | 147 |