Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019098.4 | NP_061971.3 | 15 - 16 | 1828 | 63 |
XM_011517138.2 | XP_011515440.1 | 13 - 14 | 1483 | 63 | |
GenBank mRNA | AF272900.1 | - | 15 - 16 | 1826 | 63 |
BC150601.1 | - | 15 - 16 | 1806 | 63 |