Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136193.1 | NP_001129665.1 | 4 - 5 | 1406 | 101 |
NM_001136194.1 | NP_001129666.1 | 4 - 5 | 1421 | 101 | |
NM_014929.3 | NP_055744.2 | 4 - 5 | 1313 | 101 | |
GenBank mRNA | AB023188.1 | - | 3 - 4 | 1054 | 101 |
AF223470.1 | - | 4 - 5 | 1180 | 101 | |
AK001516.1 | - | 4 - 5 | 1093 | 101 | |
AK021557.1 | - | 4 - 5 | 1207 | 101 | |
AL833877.1 | - | 4 - 5 | 1087 | 101 | |
BC001544.1 | - | 3 - 4 | 1053 | 101 |