Assay Design



Gene Transcripts

Gene Symbol
FASTKD2
Entrez Gene ID
22868
Gene Name
FAST kinase domains 2
Gene Aliases
KIAA0971
Chromosome Location
Chr.2: 206765388 - 206868284 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136193.1 NP_001129665.1 1 - 2 358 118
GenBank mRNA AF223470.1 - 1 - 2 132 118
AK021557.1 - 1 - 2 159 118
DA021045.1 - 1 - 2 170 118

View Full Product Details