Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007294.3 | NP_009225.1 | 19 - 20 | 5515 | 70 |
NM_007297.3 | NP_009228.2 | 18 - 19 | 5423 | 70 | |
NM_007298.3 | NP_009229.2 | 18 - 19 | 1990 | 70 | |
NM_007299.3 | NP_009230.2 | 19 - 20 | 2165 | 70 | |
NM_007300.3 | NP_009231.2 | 20 - 21 | 5578 | 70 | |
NR_027676.1 | 19 - 20 | 5419 | 70 | ||
GenBank mRNA | AF005068.1 | - | 19 - 20 | 5384 | 70 |
AY751490.1 | - | 18 - 19 | 5283 | 70 | |
BC072418.1 | - | 19 - 20 | 2165 | 70 | |
BC115037.1 | - | 20 - 21 | 5483 | 70 | |
DQ333386.1 | - | 4 - 5 | 357 | 70 | |
DQ363751.1 | - | 7 - 8 | 858 | 70 | |
JN686490.1 | - | 18 - 19 | 5283 | 70 | |
U14680.1 | - | 19 - 20 | 5402 | 70 | |
U64805.1 | - | 18 - 19 | 1971 | 70 | |
U68041.1 | - | 5 - 6 | 393 | 70 |