Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007294.3 | NP_009225.1 | 21 - 22 | 5638 | 96 |
NM_007297.3 | NP_009228.2 | 20 - 21 | 5546 | 96 | |
NM_007298.3 | NP_009229.2 | 20 - 21 | 2113 | 96 | |
NM_007300.3 | NP_009231.2 | 22 - 23 | 5701 | 96 | |
NR_027676.1 | 21 - 22 | 5542 | 96 | ||
GenBank mRNA | AF005068.1 | - | 21 - 22 | 5507 | 96 |
AY751490.1 | - | 20 - 21 | 5406 | 96 | |
BC115037.1 | - | 22 - 23 | 5606 | 96 | |
DQ333386.1 | - | 6 - 7 | 480 | 96 | |
DQ363751.1 | - | 9 - 10 | 981 | 96 | |
JN686490.1 | - | 20 - 21 | 5406 | 96 | |
U14680.1 | - | 21 - 22 | 5525 | 96 | |
U64805.1 | - | 20 - 21 | 2094 | 96 | |
U68041.1 | - | 7 - 8 | 516 | 96 |