Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007294.3 | NP_009225.1 | 3 - 4 | 360 | 84 |
NM_007298.3 | NP_009229.2 | 2 - 3 | 147 | 84 | |
NM_007299.3 | NP_009230.2 | 3 - 4 | 322 | 84 | |
NM_007300.3 | NP_009231.2 | 3 - 4 | 360 | 84 | |
NR_027676.1 | 3 - 4 | 289 | 84 | ||
GenBank mRNA | AF005068.1 | - | 3 - 4 | 254 | 84 |
AK308084.1 | - | 3 - 4 | 289 | 84 | |
AL701927.1 | - | 3 - 4 | 360 | 84 | |
AY354539.2 | - | 2 - 3 | 192 | 84 | |
AY751490.1 | - | 2 - 3 | 128 | 84 | |
BC030969.1 | - | 3 - 4 | 247 | 84 | |
BC072418.1 | - | 3 - 4 | 322 | 84 | |
BC085615.1 | - | 3 - 4 | 230 | 84 | |
BC106745.1 | - | 3 - 4 | 177 | 84 | |
BC115037.1 | - | 3 - 4 | 265 | 84 | |
JN686490.1 | - | 2 - 3 | 128 | 84 | |
KP272105.1 | - | - | 48 | 84 | |
U14680.1 | - | 3 - 4 | 247 | 84 | |
U64805.1 | - | 2 - 3 | 128 | 84 |