Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007298.3 | NP_009229.2 | 11 - 12 | 1069 | 129 |
NM_007299.3 | NP_009230.2 | 12 - 13 | 1244 | 129 | |
GenBank mRNA | BC072418.1 | - | 12 - 13 | 1244 | 129 |
U64805.1 | - | 11 - 12 | 1050 | 129 |