Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003675.3 | NP_003666.1 | 4 - 5 | 506 | 152 |
XM_011519754.2 | XP_011518056.1 | 5 - 6 | 631 | 152 | |
XM_017016860.1 | XP_016872349.1 | 4 - 5 | 634 | 152 | |
GenBank mRNA | AK314814.1 | - | 4 - 5 | 481 | 152 |
AU142253.1 | - | 4 - 5 | 506 | 152 | |
BC000794.1 | - | 4 - 5 | 428 | 152 | |
U51990.1 | - | 4 - 5 | 430 | 152 |