Assay Design



Gene Transcripts

Gene Symbol
DCD
Entrez Gene ID
117159
Gene Name
dermcidin
Gene Aliases
AIDD, DCD-1, DSEP, HCAP, PIF
Chromosome Location
Chr.12: 54644591 - 54648493 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300854.1 NP_001287783.1 3 - 4 385 114
NM_053283.3 NP_444513.1 3 - 4 385 114
GenBank mRNA AF144011.1 - 3 - 4 255 114
AY590150.1 - 3 - 4 249 114
BC062682.1 - 3 - 4 257 114
BC069108.1 - 3 - 4 255 114
BP313072.1 - 3 - 4 257 114
EF503687.1 - 3 - 4 385 114
EF503688.1 - 4 - 5 552 114
EF503689.1 - 3 - 4 385 114

View Full Product Details