Assay Design



Gene Transcripts

Gene Symbol
EIF4A3
Entrez Gene ID
9775
Gene Name
eukaryotic translation initiation factor 4A3
Gene Aliases
DDX48, MUK34, NMP265, NUK34, RCPS, eIF4AIII
Chromosome Location
Chr.17: 80135214 - 80147183 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014740.3 NP_055555.1 7 - 8 951 69
XM_011525522.1 XP_011523824.1 6 - 7 844 69
GenBank mRNA AK290608.1 - 7 - 8 932 69
BC003662.2 - 7 - 8 861 69
BC004386.1 - 7 - 8 882 69
BC011151.1 - 7 - 8 886 69
CR456750.1 - 7 - 8 729 69
D21853.1 - 7 - 8 943 69
X79538.1 - 7 - 8 802 69

View Full Product Details