Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015276.1 | NP_056091.1 | 8 - 9 | 1311 | 74 |
XM_005256575.2 | XP_005256632.1 | 8 - 9 | 1025 | 74 | |
GenBank mRNA | AB028986.1 | - | 8 - 9 | 1311 | 74 |
BC007196.1 | - | 1 - 2 | 53 | 74 | |
BC050994.1 | - | 1 - 2 | 468 | 74 | |
BC084550.1 | - | 1 - 2 | 468 | 74 | |
BC110499.1 | - | 8 - 9 | 1070 | 74 | |
BC126898.1 | - | 8 - 9 | 1071 | 74 | |
BX640815.1 | - | 7 - 8 | 893 | 74 |