Assay Design



Gene Transcripts

Gene Symbol
PRPF8
Entrez Gene ID
10594
Gene Name
pre-mRNA processing factor 8
Gene Aliases
HPRP8, PRP8, PRPC8, RP13, SNRNP220
Chromosome Location
Chr.17: 1650629 - 1684882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006445.3 NP_006436.3 12 - 13 1836 76
GenBank mRNA AB007510.1 - 12 - 13 1763 76
AF092565.1 - 12 - 13 1760 76
BC064370.1 - 12 - 13 1801 76

View Full Product Details