Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006445.3 | NP_006436.3 | 27 - 28 | 4453 | 68 |
GenBank mRNA | AB007510.1 | - | 27 - 28 | 4380 | 68 |
AF092565.1 | - | 27 - 28 | 4377 | 68 | |
AK296344.1 | - | 3 - 4 | 549 | 68 | |
BC064370.1 | - | 27 - 28 | 4418 | 68 |