Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006445.3 | NP_006436.3 | 34 - 35 | 5619 | 55 |
GenBank mRNA | AB007510.1 | - | 34 - 35 | 5546 | 55 |
AF092565.1 | - | 34 - 35 | 5543 | 55 | |
AK058034.1 | - | 2 - 3 | 222 | 55 | |
AK296344.1 | - | 10 - 11 | 1715 | 55 | |
AY486134.1 | - | 6 - 7 | 1010 | 55 | |
AY486135.1 | - | 6 - 7 | 1007 | 55 | |
BC064370.1 | - | 34 - 35 | 5584 | 55 |