Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005802.4 | NP_005793.2 | 1 - 2 | 193 | 72 |
GenBank mRNA | AB045732.1 | - | 1 - 2 | 120 | 72 |
AF098300.1 | - | 1 - 2 | 120 | 72 | |
AU280552.1 | - | 1 - 2 | 168 | 72 | |
BC060884.1 | - | 1 - 2 | 133 | 72 | |
DA238762.1 | - | 1 - 2 | 193 | 72 |