Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003679.4 | NP_003670.2 | 3 - 4 | 536 | 76 |
XM_005273337.1 | XP_005273394.1 | 3 - 4 | 646 | 76 | |
XM_005273338.3 | XP_005273395.1 | 4 - 5 | 889 | 76 | |
XM_017002649.1 | XP_016858138.1 | 4 - 5 | 880 | 76 | |
GenBank mRNA | AF056032.1 | - | 3 - 4 | 271 | 76 |
AK291558.1 | - | 3 - 4 | 292 | 76 | |
BC005297.1 | - | 3 - 4 | 320 | 76 | |
BP276365.1 | - | 3 - 4 | 290 | 76 | |
Y13153.1 | - | 3 - 4 | 277 | 76 |