Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003679.4 | NP_003670.2 | 7 - 8 | 932 | 148 |
XM_005273337.1 | XP_005273394.1 | 7 - 8 | 1042 | 148 | |
XM_005273338.3 | XP_005273395.1 | 8 - 9 | 1285 | 148 | |
XM_017002649.1 | XP_016858138.1 | 8 - 9 | 1276 | 148 | |
XM_017002650.1 | XP_016858139.1 | 5 - 6 | 700 | 148 | |
GenBank mRNA | AF056032.1 | - | 7 - 8 | 667 | 148 |
AK310937.1 | - | 7 - 8 | 809 | 148 | |
BC005297.1 | - | 7 - 8 | 716 | 148 | |
Y13153.1 | - | 7 - 8 | 673 | 148 |