Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024649.1 | NP_001019820.1 | 14 - 15 | 1845 | 77 |
NM_001746.3 | NP_001737.1 | 14 - 15 | 1898 | 77 | |
XM_011534664.1 | XP_011532966.1 | 14 - 15 | 2263 | 77 | |
XM_011534665.2 | XP_011532967.1 | 14 - 15 | 1871 | 77 | |
GenBank mRNA | AJ271880.1 | - | 13 - 14 | 1737 | 77 |
AK130080.1 | - | 4 - 5 | 380 | 77 | |
AK294702.1 | - | 14 - 15 | 1901 | 77 | |
AK304420.1 | - | 13 - 14 | 1780 | 77 | |
AK312334.1 | - | 14 - 15 | 1805 | 77 | |
BC003552.1 | - | 14 - 15 | 1832 | 77 | |
BC042843.1 | - | 14 - 15 | 1779 | 77 | |
L10284.1 | - | 14 - 15 | 1810 | 77 | |
L18887.1 | - | 14 - 15 | 1816 | 77 | |
M94859.1 | - | 14 - 15 | 1816 | 77 | |
M98452.1 | - | 7 - 8 | 1013 | 77 |