Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024649.1 | NP_001019820.1 | 5 - 6 | 568 | 78 |
NM_001746.3 | NP_001737.1 | 5 - 6 | 621 | 78 | |
XM_011534664.1 | XP_011532966.1 | 5 - 6 | 986 | 78 | |
XM_011534665.2 | XP_011532967.1 | 5 - 6 | 594 | 78 | |
GenBank mRNA | AJ271880.1 | - | 4 - 5 | 460 | 78 |
AK129990.1 | - | 3 - 4 | 328 | 78 | |
AK294702.1 | - | 5 - 6 | 624 | 78 | |
AK304420.1 | - | 4 - 5 | 503 | 78 | |
AK312334.1 | - | 5 - 6 | 528 | 78 | |
BC003552.1 | - | 5 - 6 | 555 | 78 | |
BC042843.1 | - | 5 - 6 | 502 | 78 | |
L10284.1 | - | 5 - 6 | 533 | 78 | |
L18887.1 | - | 5 - 6 | 539 | 78 | |
M94859.1 | - | 5 - 6 | 539 | 78 | |
S67659.1 | - | 4 - 5 | 396 | 78 |