Assay Design
Gene Transcripts
- Gene Symbol
- RGS7
- Entrez Gene ID
-
6000
- Gene Name
- regulator of G-protein signaling 7
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 240775193 - 241357374 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_002924.5 |
NP_002915.3 |
17 - 18 |
1787 |
79 |
GenBank mRNA |
BC022009.1 |
- |
17 - 18 |
1740 |
79 |
View Full Product Details