Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004939.2 | NP_004930.1 | 12 - 13 | 1131 | 89 |
GenBank mRNA | AB593031.1 | - | 12 - 13 | 884 | 89 |
AB593032.1 | - | 12 - 13 | 861 | 89 | |
AB593033.1 | - | 8 - 9 | 658 | 89 | |
AB593034.1 | - | 12 - 13 | 904 | 89 | |
AK297432.1 | - | 11 - 12 | 762 | 89 | |
AK298212.1 | - | 9 - 10 | 715 | 89 | |
AK298426.1 | - | 12 - 13 | 823 | 89 | |
AK313933.1 | - | 12 - 13 | 850 | 89 | |
BC012132.1 | - | 12 - 13 | 867 | 89 | |
BC053673.1 | - | 12 - 13 | 868 | 89 | |
EF418570.1 | - | 12 - 13 | 836 | 89 | |
EF418571.1 | - | 12 - 13 | 836 | 89 | |
X70649.1 | - | 12 - 13 | 1106 | 89 |