Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004939.2 | NP_004930.1 | 14 - 15 | 1322 | 69 |
GenBank mRNA | AB593031.1 | - | 14 - 15 | 1075 | 69 |
AB593032.1 | - | 14 - 15 | 1052 | 69 | |
AB593033.1 | - | 10 - 11 | 849 | 69 | |
AB593034.1 | - | 14 - 15 | 1095 | 69 | |
AK297432.1 | - | 13 - 14 | 953 | 69 | |
AK298212.1 | - | 11 - 12 | 906 | 69 | |
AK298426.1 | - | 14 - 15 | 1014 | 69 | |
AK313933.1 | - | 14 - 15 | 1041 | 69 | |
BC012132.1 | - | 14 - 15 | 1058 | 69 | |
BC053673.1 | - | 14 - 15 | 1059 | 69 | |
EF418570.1 | - | 14 - 15 | 1027 | 69 | |
EF418571.1 | - | 14 - 15 | 1027 | 69 | |
X70649.1 | - | 14 - 15 | 1297 | 69 |