Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004939.2 | NP_004930.1 | 20 - 21 | 2001 | 77 |
GenBank mRNA | AB593031.1 | - | 20 - 21 | 1754 | 77 |
AB593032.1 | - | 20 - 21 | 1731 | 77 | |
AB593033.1 | - | 16 - 17 | 1528 | 77 | |
AB593034.1 | - | 20 - 21 | 1774 | 77 | |
AK297432.1 | - | 19 - 20 | 1632 | 77 | |
AK298212.1 | - | 17 - 18 | 1585 | 77 | |
AK298426.1 | - | 20 - 21 | 1693 | 77 | |
AK313933.1 | - | 20 - 21 | 1720 | 77 | |
BC012132.1 | - | 20 - 21 | 1737 | 77 | |
BC053673.1 | - | 20 - 21 | 1738 | 77 | |
EF418570.1 | - | 20 - 21 | 1706 | 77 | |
EF418571.1 | - | 20 - 21 | 1706 | 77 | |
X70649.1 | - | 20 - 21 | 1976 | 77 |