Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024940.7 | NP_079216.4 | 29 - 30 | 3232 | 134 |
XM_011544661.1 | XP_011542963.1 | 29 - 30 | 3190 | 134 | |
XM_011544662.1 | XP_011542964.1 | 29 - 30 | 3190 | 134 | |
XM_011544663.2 | XP_011542965.1 | 29 - 30 | 3190 | 134 | |
XM_017013865.1 | XP_016869354.1 | 29 - 30 | 3042 | 134 | |
XM_017013866.1 | XP_016869355.1 | 27 - 28 | 4147 | 134 | |
XM_017013867.1 | XP_016869356.1 | 22 - 23 | 2580 | 134 | |
XM_017013868.1 | XP_016869357.1 | 23 - 24 | 2503 | 134 | |
GenBank mRNA | AK123607.1 | - | 29 - 30 | 3165 | 134 |
AK126249.1 | - | 4 - 5 | 351 | 134 | |
BC137175.1 | - | 29 - 30 | 3090 | 134 | |
BC137176.1 | - | 29 - 30 | 3090 | 134 | |
BX648352.1 | - | 12 - 13 | 1224 | 134 | |
CR749353.1 | - | 21 - 22 | 2373 | 134 | |
CR933718.1 | - | 17 - 18 | 1806 | 134 |