Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024940.7 | NP_079216.4 | 42 - 43 | 4544 | 70 |
XM_011544661.1 | XP_011542963.1 | 42 - 43 | 4502 | 70 | |
XM_011544662.1 | XP_011542964.1 | 42 - 43 | 4502 | 70 | |
XM_011544663.2 | XP_011542965.1 | 42 - 43 | 4502 | 70 | |
XM_017013865.1 | XP_016869354.1 | 42 - 43 | 4354 | 70 | |
XM_017013866.1 | XP_016869355.1 | 40 - 41 | 5459 | 70 | |
XM_017013867.1 | XP_016869356.1 | 35 - 36 | 3892 | 70 | |
XM_017013868.1 | XP_016869357.1 | 36 - 37 | 3815 | 70 | |
GenBank mRNA | AK126249.1 | - | 17 - 18 | 1663 | 70 |
BC041761.2 | - | 11 - 12 | 1125 | 70 | |
BC137175.1 | - | 42 - 43 | 4402 | 70 | |
BC137176.1 | - | 42 - 43 | 4402 | 70 | |
BX648352.1 | - | 26 - 27 | 2636 | 70 | |
CR933718.1 | - | 30 - 31 | 3118 | 70 |