Assay Design



Gene Transcripts

Gene Symbol
ARNTL
Entrez Gene ID
406
Gene Name
aryl hydrocarbon receptor nuclear translocator like
Gene Aliases
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
Chromosome Location
Chr.11: 13276613 - 13387268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030272.2 NP_001025443.1 6 - 7 628 131
NM_001030273.2 NP_001025444.1 7 - 8 699 202
NM_001178.5 NP_001169.3 7 - 8 683 131
NM_001297719.1 NP_001284648.1 7 - 8 683 131
NM_001297722.1 NP_001284651.1 6 - 7 628 131
NM_001297724.1 NP_001284653.1 7 - 8 692 195
XM_006718233.3 XP_006718296.1 6 - 7 570 131
XM_006718234.1 XP_006718297.1 7 - 8 636 202
XM_011520105.2 XP_011518407.2 5 - 6 734 131
XM_011520107.2 XP_011518409.2 5 - 6 729 131
XM_011520108.1 XP_011518410.1 7 - 8 676 131
XM_011520109.1 XP_011518411.1 5 - 6 431 131
XM_011520110.1 XP_011518412.1 8 - 9 691 202
XM_011520111.1 XP_011518413.1 6 - 7 506 202
XM_011520112.2 XP_011518414.1 8 - 9 691 202
XM_011520113.1 XP_011518415.1 8 - 9 691 202
XM_017017738.1 XP_016873227.1 5 - 6 732 131
XM_017017739.1 XP_016873228.1 5 - 6 728 131
XM_017017740.1 XP_016873229.1 6 - 7 570 131
XM_017017741.1 XP_016873230.1 5 - 6 431 131
XM_017017742.1 XP_016873231.1 5 - 6 431 131
XM_017017743.1 XP_016873232.1 5 - 6 431 131
XM_017017744.1 XP_016873233.1 8 - 9 691 202
XM_017017745.1 XP_016873234.1 7 - 8 636 202
XM_017017746.1 XP_016873235.1 7 - 8 646 202
XM_017017747.1 XP_016873236.1 6 - 7 506 202
XM_017017748.1 XP_016873237.1 6 - 7 563 202
GenBank mRNA AB000812.1 - 4 - 5 264 131
AB000815.1 - 5 - 6 335 202
AB000816.1 - 6 - 7 675 202
AF044288.1 - 7 - 8 593 131
AK095749.1 - 7 - 8 584 202
AK291510.1 - 6 - 7 541 131
AK315174.1 - 7 - 8 621 131
BC016674.1 - 4 - 5 227 202
BC031214.1 - 7 - 8 524 195
BC041129.1 - 7 - 8 607 131
D89722.1 - 4 - 5 240 202
U60415.1 - 6 - 7 527 131

View Full Product Details