Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018447.3 | NP_060917.1 | 5 - 6 | 878 | 70 |
XM_005265321.3 | XP_005265378.1 | 5 - 6 | 746 | 70 | |
GenBank mRNA | AF157321.1 | - | 4 - 5 | 590 | 70 |
AK172843.1 | - | 4 - 5 | 586 | 70 | |
AK222953.1 | - | 4 - 5 | 588 | 70 | |
AK312008.1 | - | 4 - 5 | 585 | 70 | |
BC022807.1 | - | 4 - 5 | 573 | 70 |