Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319952.1 | NP_001306881.1 | 7 - 8 | 889 | 64 |
NM_024526.3 | NP_078802.2 | 8 - 9 | 941 | 64 | |
NM_133181.3 | NP_573444.2 | 8 - 9 | 941 | 64 | |
NM_139053.2 | NP_620641.1 | 8 - 9 | 944 | 64 | |
XM_011542132.1 | XP_011540434.1 | 8 - 9 | 866 | 64 | |
XM_011542133.1 | XP_011540435.1 | 8 - 9 | 863 | 64 | |
XM_011542134.2 | XP_011540436.1 | 7 - 8 | 865 | 64 | |
XM_011542135.2 | XP_011540437.1 | 8 - 9 | 983 | 64 | |
XM_017002327.1 | XP_016857816.1 | 8 - 9 | 866 | 64 | |
XM_017002328.1 | XP_016857817.1 | 8 - 9 | 866 | 64 | |
XM_017002329.1 | XP_016857818.1 | 8 - 9 | 842 | 64 | |
GenBank mRNA | AK025175.1 | - | 8 - 9 | 843 | 64 |
AK225235.1 | - | 8 - 9 | 843 | 64 | |
AK290261.1 | - | 8 - 9 | 868 | 64 | |
AK292198.1 | - | 8 - 9 | 865 | 64 | |
AK307932.1 | - | 7 - 8 | 787 | 64 | |
AK308196.1 | - | 7 - 8 | 683 | 64 | |
AK310979.1 | - | 8 - 9 | 928 | 64 | |
AW937714.1 | - | 5 - 6 | 64 | ||
AY074930.1 | - | 8 - 9 | 827 | 64 | |
BC012926.1 | - | 8 - 9 | 864 | 64 |